Assay Details
Gene Symbol: CCL4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291469.1 | NP_001278398.1 | 3 | 412 | 91 | |
NM_001291470.1 | NP_001278399.1 | 2 | 814 | 91 | ||
NM_001291471.1 | NP_001278400.1 | 3 | 392 | 91 | ||
NM_001291472.1 | NP_001278401.1 | 3 | 371 | 91 | ||
NM_001291473.1 | NP_001278402.1 | 3 | 341 | 91 | ||
NM_001291474.1 | NP_001278403.1 | 2 | 297 | 91 | ||
NM_001291475.1 | NP_001278404.1 | 3 | 402 | 91 | ||
GenBank mRNA | AY766451.1 | - | 3 | 412 | 91 | |
AY766452.1 | - | 2 | 814 | 91 | ||
AY766453.1 | - | 3 | 392 | 91 | ||
AY766454.1 | - | 3 | 371 | 91 | ||
AY766455.1 | - | 3 | 341 | 91 | ||
AY766457.1 | - | 2 | 297 | 91 | ||
AY766458.1 | - | 3 | 402 | 91 | ||
BC092445.1 | - | 3 | 409 | 30416435 | 91 | |
CD366524.1 | - | 1 | 455 | 91 | ||
CD521054.1 | - | 1 | 141 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
