Assay Details
Gene Symbol: ZNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003441.2 | NP_003432.1 | 4 | 1515 | 88 | |
XM_011513562.2 | XP_011511864.1 | 3 | 2345 | 88 | ||
XM_011513563.2 | XP_011511865.1 | 2 | 2103 | 88 | ||
XM_017008591.1 | XP_016864080.1 | 4 | 2437 | 88 | ||
GenBank mRNA | BC074853.2 | - | 4 | 1375 | 30915571 | 88 |
L15309.1 | - | 4 | 1493 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
