Assay Details
| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001895.3 | NP_001886.1 | 13 | 2624 | 125 | |
| NM_177559.2 | NP_808227.1 | 14 | 2741 | 125 | ||
| NM_177560.2 | NP_808228.1 | 12 | 2414 | 125 | ||
| GenBank mRNA | BC050036.1 | - | 13 | 2513 | 5298655 | 125 |
| BC053532.1 | - | 12 | 2443 | 3908058 | 125 | |
| BC071167.1 | - | 12 | 2355 | 5772615 | 125 | |
More Information
Set Membership: |
|

CSNK2A1
Genomic Map