Assay Details
| Gene Symbol: MT1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301272.1 | NP_001288201.1 | 3 | 362 | 69 | |
| NM_005949.3 | NP_005940.1 | 3 | 320 | 69 | ||
| GenBank mRNA | BC029453.1 | - | 3 | 320 | 4041046 | 69 |
| BC108722.1 | - | 1 | 1452 | 69 | ||
| BM919862.1 | - | 3 | 285 | 69 | ||
| BQ939879.1 | - | 3 | 265 | 69 | ||
More Information
Set Membership: |
|

MT1F
Genomic Map