Assay Details
| Gene Symbol: DPY19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173812.4 | NP_776173.3 | 1 | 324 | 110 | |
| XM_017019188.1 | XP_016874677.1 | 1 | 359 | 110 | ||
| XM_017019189.1 | XP_016874678.1 | 1 | 359 | 110 | ||
| XM_017019190.1 | XP_016874679.1 | 1 | 359 | 110 | ||
| XM_017019191.1 | XP_016874680.1 | 1 | 359 | 110 | ||
| XM_017019192.1 | XP_016874681.1 | 1 | 359 | 110 | ||
| XM_017019196.1 | XP_016874685.1 | 1 | 359 | 110 | ||
| XM_017019197.1 | XP_016874686.1 | 1 | 292 | 110 | ||
| XM_017019198.1 | XP_016874687.1 | 1 | 359 | 110 | ||
| XM_017019199.1 | XP_016874688.1 | 1 | 292 | 110 | ||
| XM_017019200.1 | XP_016874689.1 | 1 | 359 | 110 | ||
| XM_017019201.1 | XP_016874690.1 | 1 | 259 | 110 | ||
| XM_017019203.1 | XP_016874692.1 | 1 | 154 | 110 | ||
| XM_017019204.1 | XP_016874693.1 | 1 | 292 | 110 | ||
| XM_017019205.1 | XP_016874694.1 | 1 | 359 | 110 | ||
| GenBank mRNA | AK057511.1 | - | 1 | 295 | 110 | |
| AK310102.1 | - | 1 | 294 | 110 | ||
| AL833344.1 | - | 1 | 110 | |||
| AY358792.1 | - | 1 | 110 | |||
| BC068442.1 | - | 1 | 5297524 | 110 | ||
| BC125215.1 | - | 1 | 214 | 110 | ||
| BC125216.1 | - | 1 | 214 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC125215.1, BC125216.1, XM_017019197.1, XM_017019199.1, XM_017019201.1, XM_017019203.1, XM_017019204.1, XM_017019205.1 transcripts.
Set Membership: |
|

DPY19L2
Genomic Map