Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278736.1 | NP_001265665.1 | 1 - 2 | 145 | 98 | |
| NM_002985.2 | NP_002976.2 | 1 - 2 | 145 | 98 | ||
| GenBank mRNA | AF043341.1 | - | 1 - 2 | 80 | 98 | |
| BC008600.1 | - | 1 - 2 | 101 | 4185200 | 98 | |
| CR980938.1 | - | 1 - 2 | 63 | 98 | ||
| M21121.1 | - | 1 - 2 | 103 | 98 | ||
More Information
Set Membership: |
|

CCL5
Genomic Map