Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006419.2 | NP_006410.1 | 3 - 4 | 274 | 73 | |
| XM_006714063.3 | XP_006714126.1 | 2 - 3 | 267 | 73 | ||
| GenBank mRNA | AF029894.1 | - | 2 - 3 | 230 | 73 | |
| AF044197.1 | - | 3 - 4 | 286 | 73 | ||
| AJ002211.1 | - | 2 - 3 | 230 | 73 | ||
| BC012589.1 | - | 2 - 3 | 246 | 4246053 | 73 | |
| BI836728.1 | - | 2 - 3 | 226 | 73 | ||
| CR407624.1 | - | 2 - 3 | 196 | 73 | ||
More Information
Set Membership: |
|

CXCL13
Genomic Map