Assay Details
| Gene Symbol: CCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031966.3 | NP_114172.1 | 3 - 4 | 613 | 96 | |
| GenBank mRNA | AM851053.1 | - | 3 - 4 | 360 | 96 | |
| BC006510.2 | - | 3 - 4 | 455 | 2963100 | 96 | |
| BT020128.1 | - | 3 - 4 | 360 | 96 | ||
| M25753.1 | - | 3 - 4 | 449 | 96 | ||
More Information
Set Membership: |
|

CCNB1
Genomic Map