Assay Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592140.2 | XP_005592197.1 | 2 - 3 | 824 | 90 | |
XM_005592141.2 | XP_005592198.1 | 2 - 3 | 809 | 90 | ||
XM_005592142.2 | XP_005592199.1 | 2 - 3 | 809 | 90 | ||
XM_015443036.1 | XP_015298522.1 | 2 - 3 | 824 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
