Assay Details
| Gene Symbol: HSD17B8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285410.1 | NP_001272339.1 | 7 - 8 | 693 | 90 | |
| GenBank mRNA | DQ228170.1 | - | 7 - 8 | 693 | 90 | |
More Information
Set Membership: |
|

HSD17B8
Genomic Map