Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569431.2 | XP_005569488.1 | 2 - 3 | 307 | 77 | |
XM_005569432.2 | XP_005569489.1 | 2 - 3 | 304 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
