Assay Details
| Gene Symbol: UCMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564642.2 | XP_005564699.1 | 4 - 5 | 412 | 89 | |
| XM_015455557.1 | XP_015311043.1 | 3 - 4 | 316 | 89 | ||
More Information
Set Membership: |
|

UCMA
Genomic Map