Assay Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553730.2 | XP_005553787.1 | 5 - 6 | 814 | 74 | |
XM_005553731.2 | XP_005553788.1 | 5 - 6 | 573 | 74 | ||
XM_005553732.2 | XP_005553789.1 | 5 - 6 | 722 | 74 | ||
XM_015449749.1 | XP_015305235.1 | 5 - 6 | 576 | 74 | ||
XM_015449750.1 | XP_015305236.1 | 6 - 7 | 995 | 74 | ||
XM_015449751.1 | XP_015305237.1 | 4 - 5 | 508 | 74 | ||
XM_015449752.1 | XP_015305238.1 | 4 - 5 | 402 | 74 | ||
XM_015449753.1 | XP_015305239.1 | 3 - 4 | 325 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015449749.1, XM_015449750.1, XM_015449751.1, XM_015449752.1, XM_015449753.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
