Assay Details
Gene Symbol: CAVIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578812.2 | XP_005578869.1 | 1 - 2 | 1229 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
