Assay Details
| Gene Symbol: OTUD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319544.1 | NP_001306473.1 | 2 - 3 | 494 | 94 | |
| XM_005544570.2 | XP_005544627.2 | 2 - 3 | 488 | 94 | ||
| GenBank mRNA | AB173508.1 | - | 2 - 3 | 494 | 94 | |
More Information
Set Membership: |
|

OTUD3
Genomic Map