Assay Details
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545549.2 | XP_005545606.1 | 3 - 4 | 735 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
