Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567311.2 | XP_005567368.1 | 2 - 3 | 399 | 61 | |
| XM_005567312.2 | XP_005567369.1 | 2 - 3 | 258 | 61 | ||
More Information
Set Membership: |
|

MAFF
Genomic Map