Assay Details
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555321.2 | XP_005555378.1 | 5 - 6 | 1770 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
