Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286228.1 | NP_001273157.1 | 3 | 616 | 131 | |
GenBank mRNA | DK584637.1 | - | 3 | 555 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
