Assay Details
| Gene Symbol: PRSS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442393.1 | XP_015297879.1 | 5 - 6 | 1970 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS41| Gene Symbol: PRSS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442393.1 | XP_015297879.1 | 5 - 6 | 1970 | 70 | |
Set Membership: |
|