Assay Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581025.2 | XP_005581082.1 | 3 - 4 | 444 | 99 | |
XM_005581026.2 | XP_005581083.1 | 3 - 4 | 289 | 99 | ||
XM_005581027.2 | XP_005581084.1 | 3 - 4 | 353 | 99 | ||
XM_005581028.2 | XP_005581085.1 | 3 - 4 | 315 | 99 | ||
XM_005581029.2 | XP_005581086.1 | 3 - 4 | 411 | 99 | ||
XM_005581030.2 | XP_005581087.1 | 3 - 4 | 381 | 99 | ||
XM_005581031.1 | XP_005581088.1 | 3 - 4 | 437 | 99 | ||
XM_015436851.1 | XP_015292337.1 | 3 - 4 | 319 | 99 | ||
XM_015436852.1 | XP_015292338.1 | 2 - 3 | 476 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
