Assay Details
| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567948.2 | XP_005568005.1 | 3 - 4 | 536 | 62 | |
| XM_005567949.2 | XP_005568006.1 | 3 - 4 | 536 | 62 | ||
More Information
Set Membership: |
|

RANBP1
Genomic Map