Assay Details
| Gene Symbol: TCAIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546826.2 | XP_005546883.1 | 7 - 8 | 1224 | 81 | |
| XM_005546827.2 | XP_005546884.1 | 7 - 8 | 1038 | 81 | ||
| XM_005546828.2 | XP_005546885.1 | 6 - 7 | 1101 | 81 | ||
| XM_005546830.1 | XP_005546887.1 | 5 - 6 | 648 | 81 | ||
| XM_015445732.1 | XP_015301218.1 | 7 - 8 | 857 | 81 | ||
| XM_015445733.1 | XP_015301219.1 | 6 - 7 | 951 | 81 | ||
| XM_015445734.1 | XP_015301220.1 | 8 - 9 | 942 | 81 | ||
| XM_015445735.1 | XP_015301221.1 | 7 - 8 | 1145 | 81 | ||
| XM_015445737.1 | XP_015301223.1 | 7 - 8 | 962 | 81 | ||
| XM_015445738.1 | XP_015301224.1 | 8 - 9 | 1016 | 81 | ||
| XM_015445739.1 | XP_015301225.1 | 5 - 6 | 1103 | 81 | ||
| XM_015445740.1 | XP_015301226.1 | 6 - 7 | 1102 | 81 | ||
| XM_015445741.1 | XP_015301227.1 | 6 - 7 | 890 | 81 | ||
| XM_015445742.1 | XP_015301228.1 | 6 - 7 | 817 | 81 | ||
| XM_015445743.1 | XP_015301229.1 | 4 - 5 | 493 | 81 | ||
| GenBank mRNA | AB168216.1 | - | 7 - 8 | 950 | 81 | |
| AB174127.1 | - | 6 - 7 | 1161 | 81 | ||
More Information
Set Membership: |
|

TCAIM
Genomic Map