Assay Details
| Gene Symbol: POP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563773.2 | XP_005563830.1 | 10 - 11 | 1576 | 73 | |
| XM_015455060.1 | XP_015310546.1 | 10 - 11 | 1581 | 73 | ||
More Information
Set Membership: |
|

POP1
Genomic Map