Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283339.1 | NP_001270268.1 | 4 - 5 | 786 | 69 | |
| GenBank mRNA | AB168325.1 | - | 4 - 5 | 786 | 69 | |
More Information
Set Membership: |
|

ZNF410
Genomic Map