Assay Details
| Gene Symbol: MAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591813.2 | XP_005591870.2 | 2 - 3 | 632 | 65 | |
| XM_015442418.1 | XP_015297904.1 | 2 - 3 | 632 | 65 | ||
More Information
Set Membership: |
|

MAZ
Genomic Map