Assay Details
| Gene Symbol: RIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553891.2 | XP_005553948.1 | 2 - 3 | 596 | 76 | |
| XM_005553892.2 | XP_005553949.1 | 2 - 3 | 593 | 76 | ||
| XM_015449829.1 | XP_015305315.1 | 2 - 3 | 938 | 76 | ||
| XM_015449836.1 | XP_015305322.1 | 1 - 2 | 193 | 76 | ||
| XM_015449838.1 | XP_015305324.1 | 1 - 2 | 193 | 76 | ||
| XM_015449839.1 | XP_015305325.1 | 1 - 2 | 193 | 76 | ||
| XM_015449843.1 | XP_015305329.1 | 1 - 2 | 2342 | 76 | ||
More Information
Set Membership: |
|

RIPOR2
Genomic Map