Assay Details
| Gene Symbol: FRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553039.2 | XP_005553096.1 | 3 - 4 | 1843 | 79 | |
| XM_005553040.2 | XP_005553097.1 | 3 - 4 | 1837 | 79 | ||
| XM_005553042.1 | XP_005553099.1 | 4 - 5 | 825 | 79 | ||
| XM_015449413.1 | XP_015304899.1 | 4 - 5 | 1602 | 79 | ||
| XM_015449414.1 | XP_015304900.1 | 4 - 5 | 1208 | 79 | ||
More Information
Set Membership: |
|

FRS3
Genomic Map