Assay Details
| Gene Symbol: C11H12orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572421.2 | XP_005572478.1 | 3 - 4 | 309 | 66 | |
| XM_005572422.2 | XP_005572479.1 | 3 - 4 | 309 | 66 | ||
| XM_005572423.2 | XP_005572480.1 | 3 - 4 | 396 | 66 | ||
More Information
Set Membership: |
|

C11H12orf43
Genomic Map