Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283520.1 | NP_001270449.1 | 4 - 5 | 530 | 81 | |
| GenBank mRNA | AB179272.1 | - | 4 - 5 | 530 | 81 | |
More Information
Set Membership: |
|

RNF114
Genomic Map