Assay Details
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284139.1 | NP_001271068.1 | 839 | 57 | ||
GenBank mRNA | AB170104.1 | - | 839 | 57 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
