Assay Details
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285236.1 | NP_001272165.1 | 9 - 10 | 1257 | 63 | |
XM_005593053.2 | XP_005593110.1 | 9 - 10 | 1405 | 63 | ||
GenBank mRNA | AB179464.1 | - | 9 - 10 | 1257 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
