Assay Details
| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581274.2 | XP_005581331.1 | 8 - 9 | 878 | 83 | |
| XM_005581275.2 | XP_005581332.1 | 7 - 8 | 830 | 83 | ||
More Information
Set Membership: |
|

GRHPR
Genomic Map