Assay Details
Gene Symbol: BRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284122.1 | NP_001271051.1 | 2 - 3 | 323 | 97 | |
XM_005563056.2 | XP_005563113.1 | 2 - 3 | 339 | 97 | ||
GenBank mRNA | AB179450.1 | - | 2 - 3 | 323 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
