Assay Details
Gene Symbol: HTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558201.2 | XP_005558258.1 | 5 - 6 | 1216 | 88 | |
XM_015452110.1 | XP_015307596.1 | 5 - 6 | 715 | 88 | ||
XM_015452111.1 | XP_015307597.1 | 5 - 6 | 993 | 88 | ||
XM_015452112.1 | XP_015307598.1 | 5 - 6 | 713 | 88 | ||
XM_015452114.1 | XP_015307600.1 | 5 - 6 | 714 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
