Assay Details
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551128.2 | XP_005551185.1 | 2 - 3 | 1867 | 79 | |
XM_015448263.1 | XP_015303749.1 | 2 - 3 | 1797 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
