Assay Details
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579375.2 | XP_005579432.1 | 17 - 18 | 2158 | 110 | |
XM_005579376.2 | XP_005579433.1 | 17 - 18 | 2132 | 110 | ||
XM_005579378.2 | XP_005579435.1 | 17 - 18 | 2155 | 110 | ||
XM_015435613.1 | XP_015291099.1 | 16 - 17 | 2029 | 110 | ||
XM_015435614.1 | XP_015291100.1 | 14 - 15 | 1684 | 110 | ||
GenBank mRNA | AB125181.1 | - | 17 - 18 | 2156 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
