Assay Details
| Gene Symbol: NKX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561100.2 | XP_005561157.1 | 2 - 3 | 729 | 72 | |
| XM_015453718.1 | XP_015309204.1 | 3 - 4 | 874 | 72 | ||
More Information
Set Membership: |
|

NKX2-1
Genomic Map