Assay Details
| Gene Symbol: C18H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586800.2 | XP_005586857.1 | 1 - 2 | 209 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005586800.2 transcripts.
Set Membership: |
|

C18H18orf25
Genomic Map