Assay Details
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283994.1 | NP_001270923.1 | 8 - 9 | 1067 | 98 | |
XM_005547878.2 | XP_005547935.1 | 8 - 9 | 833 | 98 | ||
XM_005547881.2 | XP_005547938.1 | 7 - 8 | 716 | 98 | ||
XM_015445354.1 | XP_015300840.1 | 8 - 9 | 1186 | 98 | ||
GenBank mRNA | AB052134.1 | - | 8 - 9 | 1067 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
