Assay Details
| Gene Symbol: LOC102140649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553357.2 | XP_005553414.1 | 3 - 4 | 1731 | 77 | |
| XM_005553358.2 | XP_005553415.1 | 3 - 4 | 1732 | 77 | ||
More Information
Set Membership: |
|

LOC102140649
Genomic Map