Assay Details
Gene Symbol: MESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283282.1 | NP_001270211.1 | 2 - 3 | 448 | 69 | |
GenBank mRNA | AB170087.1 | - | 2 - 3 | 449 | 69 | |
AB170880.1 | - | 2 - 3 | 450 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
