Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541267.2 | XP_005541324.1 | 2 - 3 | 376 | 74 | |
| GenBank mRNA | AB169572.1 | - | 2 - 3 | 315 | 74 | |
More Information
Set Membership: |
|

PEA15
Genomic Map