Assay Details
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287705.1 | NP_001274634.1 | 1 - 2 | 174 | 78 | |
GenBank mRNA | AB172554.1 | - | 1 - 2 | 174 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
