Assay Details
Gene Symbol: HSD17B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283105.1 | NP_001270034.1 | 4 - 5 | 478 | 93 | |
GenBank mRNA | AB169576.1 | - | 4 - 5 | 478 | 93 | |
AB170661.1 | - | 4 - 5 | 478 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
