Assay Details
Gene Symbol: KIF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593863.2 | XP_005593920.1 | 29 - 30 | 3520 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
