Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553431.2 | XP_005553488.1 | 7 - 8 | 2130 | 80 | |
XM_005553432.2 | XP_005553489.1 | 7 - 8 | 2676 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
