Assay Details
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589787.2 | XP_005589844.1 | 2 - 3 | 885 | 70 | |
XM_005589788.2 | XP_005589845.1 | 2 - 3 | 882 | 70 | ||
XM_005589791.2 | XP_005589848.1 | 2 - 3 | 881 | 70 | ||
XM_005589792.2 | XP_005589849.1 | 2 - 3 | 877 | 70 | ||
GenBank mRNA | AB171927.1 | - | 2 - 3 | 873 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
