Assay Details
| Gene Symbol: GPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544278.2 | XP_005544335.1 | 1 - 2 | 114 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005544278.2 transcripts.
Set Membership: |
|

GPR3
Genomic Map