Assay Details
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570775.2 | XP_005570832.1 | 5 - 6 | 2597 | 78 | |
XM_005570777.2 | XP_005570834.1 | 5 - 6 | 2597 | 78 | ||
XM_015430886.1 | XP_015286372.1 | 5 - 6 | 2598 | 78 | ||
XM_015430887.1 | XP_015286373.1 | 5 - 6 | 2346 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
